missing translation for 'onlineSavingsMsg'
Learn More

Desmedipham, TRC

Product Code. 30368316
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30368316 5 g 5g
30356912 25 g 25g
30354607 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368316 Supplier Toronto Research Chemicals Supplier No. TRCD2821005G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13684-56-5
Molecular Formula C16 H16 N2 O4
Molecular Weight (g/mol) 300.31
Synonym Carbamic acid, [3-[[(phenylamino)carbonyl]oxy]phenyl]-, ethyl ester (9CI), Carbanilic acid, m-hydroxy-, ethyl ester, carbanilate (ester) (8CI), 3-Ethoxycarbonylaminophenyl N-phenylcarbamate, Betanex, DMP Stefes, Desmedipham, Destor, EP 475, EP 475 (pesticide), Ethyl N-[3-(N-phenylcarbamoyloxy)phenyl]carbamate, Ethyl m-hydroxycarbanilate carbanilate, SN 38107, Synbetan D
IUPAC Name [3-(ethoxycarbonylamino)phenyl] N-phenylcarbamate
SMILES CCOC(=O)Nc1cccc(OC(=O)Nc2ccccc2)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H16N2O4/c1-2-21-15(19)18-13-9-6-10-14(11-13)22-16(20)17-12-7-4-3-5-8-12/h3-11H,2H2,1H3,(H,17,20)(H,18,19)
Formula Weight 300.111
Chemical Name or Material Desmedipham
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.