missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Chlorpheniramine Maleate Salt, TRC

Product Code. 18040440
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18040440 5 mg 5mg
18000930 1 mg 1mg
18020230 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18040440 Supplier Toronto Research Chemicals Supplier No. TRCD2915505MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22630-25-7
Molecular Formula C15 H17 Cl N2 . C4 H4 O4
Molecular Weight (g/mol) 376.83
Synonym (3RS)-3-(4-Chloro-phenyl)-N-methyl-3-(pyridin-2-yl)propan-1-amine Maleate, Chlorphenamine Maleate Imp. C (EP) as Maleate, Desmethylchlorphenamine Maleate, Pyridine, 2-[p-chloro-α-[2-(methylamino)ethyl]benzyl]-, maleate (1:1) (8CI), 2-[p-Chloro-α-(2-methylaminoethyl)benzyl]pyridine maleate
IUPAC Name (Z)-but-2-enedioic acid;3-(4-chlorophenyl)-N-methyl-3-pyridin-2-ylpropan-1-amine
SMILES CNCCC(c1ccc(Cl)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H17ClN2.C4H4O4/c1-17-11-9-14(15-4-2-3-10-18-15)12-5-7-13(16)8-6-12;5-3(6)1-2-4(7)8/h2-8,10,14,17H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formula Weight 376.119
Chemical Name or Material Desmethylchlorphenamine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.