missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Thiosildenafil, TRC

Product Code. 30363216
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363216 25 mg 25mg
30354472 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30363216 Supplier Toronto Research Chemicals Supplier No. TRCD29435025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479073-86-4
Molecular Formula C21 H28 N6 O3 S2
Molecular Weight (g/mol) 476.62
Synonym Piperazine, 1-[[3-(4,7-dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]- (9CI), 5-[2-Ethoxy-5-(1-piperazinylsulfonyl)phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, Thiosildenafil Desmethyl
IUPAC Name 5-(2-ethoxy-5-piperazin-1-ylsulfonylphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidine-7-thione
SMILES CCCc1nn(C)c2C(=S)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCNCC4

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H28N6O3S2/c1-4-6-16-18-19(26(3)25-16)21(31)24-20(23-18)15-13-14(7-8-17(15)30-5-2)32(28,29)27-11-9-22-10-12-27/h7-8,13,22H,4-6,9-12H2,1-3H3,(H,23,24,31)
Formula Weight 476.1664
Chemical Name or Material Desmethyl Thiosildenafil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.