missing translation for 'onlineSavingsMsg'
Learn More

Desmethylmaprotiline Hydrochloride, TRC

Product Code. 30040632
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040632 10 mg 10mg
30009609 5 mg 5mg
18043240 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040632 Supplier Toronto Research Chemicals Supplier No. TRCD29179010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92202-51-2
Molecular Formula C19 H21 N . Cl H
Molecular Weight (g/mol) 299.84
Synonym 9,10-Ethanoanthracene-9(10H)-propanamine, hydrochloride (9CI), Demethylmaprotiline hydrochloride, Desmethylmaprotiline hydrochloride, N-Desmethylmaprotiline hydrochloride, Normaprotiline hydrochloride, 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine hydrochloride, Maprotiline-desmethyl Hydrochloride
IUPAC Name 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine hydrochloride
SMILES Cl.NCCCC12CCC(c3ccccc13)c4ccccc24

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H21N.ClH/c20-13-5-11-19-12-10-14(15-6-1-3-8-17(15)19)16-7-2-4-9-18(16)19;/h1-4,6-9,14H,5,10-13,20H2;1H
Formula Weight 299.1441
Percent Purity >95%
Chemical Name or Material 3-(9,10-Ethanoanthracen-9(10H)-yl)propan-1-amine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.