missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Desmocollin 2 Monoclonal Antibody (06)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530495
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human DSC2.
This gene encodes a member of the desmocollin protein subfamily. Desmocollins, along with desmogleins, are cadherin-like transmembrane glycoproteins that are major components of the desmosome. Desmosomes are cell-cell junctions that help resist shearing forces and are found in high concentrations in cells subject to mechanical stress. This gene is found in a cluster with other desmocollin family members on chromosome 18. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11, and reduced protein expression has been described in several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications
| Desmocollin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q02487 | |
| DSC2 | |
| Recombinant Human DSC2 Protein (Met1-Arg684). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 06 | |
| Unconjugated | |
| DSC2 | |
| ARVC10; ARVD10; ARVD11; AW228162; Cadherin family member 2; CDHF2; CDHF5; CDHF6; CMD1BB; desmocollin 2; desmocollin-2; Desmocollin-3; desmoglein-2; desmoglein-3; Desmosomal glycoprotein II; desmosomal glycoprotein II/III; Desmosomal glycoprotein III; DG2; DGII/III; DSC2; Dsc2a; Dsc2b; DSC3; Epithelial type 2 desmocollin; HDGC; PVA | |
| Mouse | |
| Protein A | |
| RUO | |
| 1824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction