missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoplakin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5115145
Additional Details : Weight : 0.01000kg
Description
Antibody detects endogenous levels of total DSP.
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.Specifications
Desmoplakin | |
Polyclonal | |
Unconjugated | |
DSP | |
2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
Rabbit | |
Affinity chromatography | |
RUO | |
109620, 1832 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
E9Q557, P15924 | |
DSP | |
A synthesized peptide derived from human DSP(Accession P15924), corresponding to amino acid residues S2792-R2842. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |