Learn More
Desmoplakin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561103
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: NHNKVIETNR ENDKQETWML MELQKIRRQI EHCEGRMTLK NLPLADQGSS HHITVKINEL KSVQNDSQAI AEVLNQLKDM LANFRGSEKY CYLQNEVF Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.Specifications
Desmoplakin | |
Polyclonal | |
Unconjugated | |
DSP | |
2300002E22Rik; 250/210 kDa paraneoplastic pemphigus antigen; 5730453H04Rik; AA407887; AA407888; AW109828; DCWHKTA; dermatan sulfate proteoglycan 3; desmoplakin; desmoplakin I/II; Desmoplakin II; desmoplakin; LOW QUALITY PROTEIN: desmoplakin; Desmoplakin-I; desmosomal cytoskeletal connector molecule; DP; Dsp; dspg3; Oculoglycan; Opt; Optc; optcl; Opticin; rul; SLRP; SLRP protein; small leucine-rich repeat protein; small leucine-rich repeat protein (SLRP); wu:fb81a05; zgc:101047 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1832 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P15924 | |
DSP | |
Recombinant protein corresponding to Human Desmoplakin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only