missing translation for 'onlineSavingsMsg'
Learn More

Desmopressin Acetate, TRC

Product Code. 17944685
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17944685 10 mg 10mg
17942555 50 mg 50mg
17921865 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944685 Supplier Toronto Research Chemicals Supplier No. TRCD29679010MG
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62288-83-9
Synonym Vasopressin, 1-(3-mercaptopropanoic acid)-8-D-arginine-, monoacetate (salt) (9CI)∣1,2-Dithia-5,8,11,14,17-pentaazacycloeicosane, cyclic peptide deriv.∣1-(3-Mercaptopropionic acid)-8-D-arginine vasopressin monoacetate∣Desmopressin acetate∣Emosint∣Minirine∣Octostim∣Stimate

Specifications

Format Neat
Molecular Weight (g/mol) 1129.27
Product Type API
Quantity 10 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.