Learn More
Desogestrel, LoGiCal
Product Code. 30461462
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461462 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461462 Supplier LoGiCal Supplier No. LGCFOR1024.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desogestrel
CAS 54024-22-5
Analyte or Component Names Desogestrel
Shipping Condition Room Temperature
Molecular Formula C22 H30 O
Certifications/Compliance ISO 9001
Synonym Desogestrel, (+)-Desogestrel, Cerazette, Cyclosa, Dicromil, Marvelon 150/320, Org 2969, 13-Ethyl-11-methylidene-18,19-dinor-17alpha-pregn-4-en-20-yn-17-ol, (+)-13beta-Ethyl-17alpha-ethynyl-11-methylenegon-4-en-17beta-ol, 13-Ethyl-11-methylene-18,19-dinor-17alpha-4-pregnen-20-yn-17-ol, 17-Ethynyl-18-methyl-11-methyleneestr-4-en-17beta-ol
InChI Formula InChI=1S/C22H30O/c1-4-21-14-15(3)20-17-9-7-6-8-16(17)10-11-18(20)19(21)12-13-22(21,23)5-2/h2,8,17-20,23H,3-4,6-7,9-14H2,1H3/t17-,18-,19-,20+,21-,22-/m0/s1
Solution Type Neat
SMILES CC[C@]12CC(=C)[C@H]3[C@@H](CCC4=CCCC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
IUPAC Name (8S,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-11-methylidene-1,2,3,6,7,8,9,10,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-ol
Industry Type Clinical Testing
Molecular Weight (g/mol) 310.47
Quantity 10 mg
Formula Weight 310.2297 g/mol