missing translation for 'onlineSavingsMsg'
Learn More

Desoxymetasone, TRC

Product Code. 17943715
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943715 10 mg 10mg
17941865 5 mg 5mg
17973755 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943715 Supplier Toronto Research Chemicals Supplier No. TRCD29697010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 382-67-2
Molecular Formula C22 H29 F O4
Molecular Weight (g/mol) 376.46
Synonym 9-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, Dexamethason Impurity F, : Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,21-dihydroxy-16α-methyl- (7CI,8CI), 17-Deoxydexamethasone, 17-Deoxymethasone, 17-Desoximethasone, 9-Fluoro-16α-methyl-δ1-corticosterone, 9α-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione, A 41-304, A 41304, Deoxymethasone, Desoximetasone, Desoximethasone, Desoxymetasone, Desoxymethasone, Esperson, HOE 304, R 2113, Stidex, Topicort, Topicorte, Topisolon
IUPAC Name (8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@H]1C(=O)CO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H29FO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,23)18(27)10-20(16,2)19(12)17(26)11-24/h6-7,9,12,15-16,18-19,24,27H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,18+,19-,20+,21+,22+/m1/s1
Formula Weight 376.205
Percent Purity >95%
Chemical Name or Material Desoximetasone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.