missing translation for 'onlineSavingsMsg'
Learn More

Despentamino Pentazido Tobramycin Tetrabenzyl Ether 6' '-tert-Butyldimethysilyl Ether, TRC

Product Code. 30363143
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363143 5 mg 5mg
30357291 1 mg 1mg
30356981 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363143 Supplier Toronto Research Chemicals Supplier No. TRCD2976105MG
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Despentamino Pentazido Tobramycin Tetrabenzyl Ether 6''-tert-Butyldimethysilyl Ether
Analyte or Component Names Despentamino Pentazido Tobramycin Tetrabenzyl Ether 6''-tert-Butyldimethysilyl Ether
Shipping Condition Room Temperature
Molecular Formula C52H65N15O9Si
InChI Formula InChI=1S/C52H65N15O9Si/c1-52(2,3)77(4,5)72-33-43-47(69-30-35-20-12-7-13-21-35)44(62-67-57)48(70-31-36-22-14-8-15-23-36)51(74-43)76-46-39(60-65-55)26-38(59-64-54)45(49(46)71-32-37-24-16-9-17-25-37)75-50-40(61-66-56)27-41(42(73-50)28-58-63-53)68-29-34-18-10-6-11-19-34/h6-25,38-51H,26-33H2,1-5H3/t38?,39?,40?,41?,42-,43-,44?,45-,46+,47?,48?,49?,50-,51-/m1/s1
Solution Type Neat
SMILES [N-]=[N+]=N[C@H]1[C@@H](O[C@@H]([C@@H]2OCC3=CC=CC=C3)[C@@H](N=[N+]=[N-])C[C@@H](N=[N+]=[N-])[C@@H]2O[C@@H]4[C@H](OCC5=CC=CC=C5)[C@@H](N=[N+]=[N-])[C@H](OCC6=CC=CC=C6)[C@@H](CO[Si](C)(C)C(C)(C)C)O4)O[C@H](CN=[N+]=[N-])[C@@H](OCC7=CC=CC=C7)C1
IUPAC Name (((2R,3S,4S,5R,6S)-4-azido-3,5-bis(benzyloxy)-6-(((1S,2S,3R,4S,6R)-4,6-diazido-3-(((2R,3R,5S,6R)-3-azido-6-(azidomethyl)-5-(benzyloxy)tetrahydro-2H-pyran-2-yl)oxy)-2-(benzyloxy)cyclohexyl)oxy)tetrahydro-2H-pyran-2-yl)methoxy)(tert-butyl)dimethylsilane
Industry Type Pharmaceutical
Molecular Weight (g/mol) 1072.25
Quantity 5 mg
Formula Weight 1071.89
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.