missing translation for 'onlineSavingsMsg'
Learn More

Deterenol Hydrochloride, TRC

Product Code. 30135608
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135608 25 mg 25mg
17933175 10 mg 10mg
17943165 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135608 Supplier Toronto Research Chemicals Supplier No. TRCD29795025MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23239-36-3
Molecular Formula C11H18ClNO2
Molecular Weight (g/mol) 231.72
Synonym Benzenemethanol, 4-hydroxy-α-[[(1-methylethyl)amino]methyl]-, hydrochloride (9CI), Benzenemethanol, 4-hydroxy-α-[[(1-methylethyl)amino]methyl]-, hydrochloride, (±)-, Benzyl alcohol, p-hydroxy-α-[(isopropylamino)methyl]-, hydrochloride, (±)- (8CI), (±)-1-(4-Hydroxyphenyl)-2-isopropylaminoethanol hydrochloride, Detenerol hydrochloride, Deterenol hydrochloride, dl-1-(4-Hydroxyphenyl)-2-(isopropylamino)ethanol hydrochloride, Benzenemethanol, 4-hydroxy-α-[[(1-methylethyl)amino]methyl]-, hydrochloride (1:1)
IUPAC Name 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]phenol;hydrochloride
SMILES Cl.CC(C)NCC(O)c1ccc(O)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H17NO2.ClH/c1-8(2)12-7-11(14)9-3-5-10(13)6-4-9;/h3-6,8,11-14H,7H2,1-2H3;1H
Formula Weight 231.1026
Chemical Name or Material Deterenol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.