Learn More
Taylor Wharton™ LD Series Cryogenic Storage Dewar
Product Code. 11778519
Change view
Click to view available options
Capacity (Metric):
4 L
25 L
35 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11778519 25 L Each
11758519 4 L Each
11788519 35 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11778519 Supplier Taylor Wharton™ Supplier No. 25LDB
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
With sealed vacuum space between the inner and outer shells to provide superior performance and insulation provides maximum holding times.
Specifications
Type Cryogenic Storage Dewar
Capacity (English) 6.6 gal.
Capacity (Metric) 25 L
Static Holding Time 109 Days