Learn More
Dexamethasone Sodium Phosphate, TRC
Product Code. 18053450
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18053450 1 g 1g
30040756 100 mg 100mg
18030240 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18053450 Supplier Toronto Research Chemicals Supplier No. TRCD2988981G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2392-39-4
Molecular Formula C22 H28 F O8 P . 2 Na
Molecular Weight (g/mol) 516.4
Synonym Dexamethasone Sodium Phosphate, (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione sodium salt (1:2), (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-(phosphonooxy)pregna-1,4-diene-3,20-dione disodium salt, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-(dihydrogen phosphate) disodium salt, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregna-1,4-diene-3,20-dione 21-phosphate disodium salt, Ak-Dex, Baldex, Colvasone, Corson, Corson (pharmaceutical), Dalalone, Decadron phosphate, Decdan, Dexabene, Dexacort, Dexadreson, Dexagel, Dexagro, Dexamethasone 21-(disodium phosphate), Dexamethasone 21-phosphate disodium salt, Dexamethasone disodium phosphate, Dexamethasone phosphate disodium salt, Dexamethasone phosphate sodium salt, Dexamethasone sodium phosphate, Dexamethazone sodium phosphate, Dezone, Disodium dexamethasone 21-monophosphate, Disodium dexamethasone 21-phosphate, Disodium dexamethasone phosphate, Maxidex Ointment, Megacort, Onadron, Orgadrone, Sodium dexamethasone phosphate, Soldesam, Solu-Deca, Solu-Decadron, Spersadex, Turbinaire
IUPAC Name disodium;[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] phosphate
SMILES [Na+].[Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COP(=O)([O-])[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15+,16+,17+,19+,20+,21+,22+;;/m1../s1
Formula Weight 516.1301
Chemical Name or Material Dexamethasone disodium phosphate