missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dexrazoxane hydrochloride
GREENER_CHOICE

Product Code. 18702744
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18702744

Brand: Tocris Bioscience™ 3857/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Topoisomerase II inhibitor

Topoisomerase II inhibitor and intracellular ion chelator. Bridges and stabilizes an interface between two ATPase promoters to inhibit topoisomerase II activity. Cardioprotective when co-administered with doxorubicin; decreases formation of reactive oxygen species (ROS) and activates the PI3K/Akt survival pathway.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1263283-43-7
Molecular Formula C11H17ClN4O4

Specifications

Quantity 10 mg
Formula Weight 304.73
Purity >98%
Molecular Weight (g/mol) 304.731
Chemical Name or Material Dexrazoxane hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.