missing translation for 'onlineSavingsMsg'
Learn More

Dextrin, precipitated by alcohol

Product Code. 11477697
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11477697

Brand: Thermo Scientific Alfa Aesar A15717.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9004-53-9 | C18H32O16 | 504.438 g/mol

Dextrin is used as adhesive in the manufacture of gummed tapes, textiles and paper. It is used in producing nutritional products. It is also used as a absorbents, binding agents and viscosity controlling agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-53-9
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00081554
InChI Key FYGDTMLNYKFZSV-MRCIVHHJSA-N
Synonym dextrin, dextrins, fortodex, dextrid, corn dextrin, dextrina bianca, british gum, crystal gum, dextrin 3, caloreen
PubChem CID 62698
IUPAC Name (3R,4S,5S,6R)-2-[(2R,3S,4R,5R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Merck Index 14,2953
Solubility Information Soluble in hot water (0.1g/ml).
Quantity 100 g
Sensitivity Hygroscopic
Chemical Name or Material Dextrin, precipitated by alcohol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.