missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DFF40/CAD Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
610.00€
Specifications
| Antigen | DFF40/CAD |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 610 SE |
| Form | Purified |
Description
DFF40/CAD Polyclonal antibody specifically detects DFF40/CAD in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| DFF40/CAD | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CADCaspase-activated nuclease, Caspase-activated deoxyribonuclease, Caspase-activated DNase, CPANDNA fragmentation factor 40 kDa subunit, DFF-40DFF40DFF2DNA fragmentation factor subunit beta, DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase), EC 3.- | |
| Antibody was raised against a peptide corresponding to 18 amino acids near the center of murine CAD. The immunogen is located within amino acids 130 - 180 of DFF40. Amino Acid Squence: GLESRFRNKSGYLRYSC | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 610 SE | |
| Rabbit | |
| Apoptosis | |
| 50mM Sodium Borate | |
| 1677 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title