missing translation for 'onlineSavingsMsg'
Learn More

DHX9 Mouse anti-Human, Mouse, Rat, CoraLite 488, Clone: 1B12C10, Proteintech

Mouse Monoclonal Antibody

145.00€ - 450.00€

Specifications

Antigen DHX9
Clone 1B12C10
Applications Immunofluorescence, Flow Cytometry
Classification Monoclonal
Conjugate CoraLite 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17701309
View Documents
Proteintech
CL488-67153-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17649876
View Documents
Proteintech
CL488-67153-100UL
100 μL
450.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DHX9 Monoclonal antibody apecifically detetects DHX9 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifications

Specifications

DHX9
Immunofluorescence, Flow Cytometry
CoraLite 488
Mouse
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
1660
DHX9 fusion protein Ag12104
Primary
Store at -20°C. Avoid exposure to light.
1B12C10
Monoclonal
Liquid
Human, Mouse, Rat
BC014246
DHX9
IgG1
Protein G purification
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.