missing translation for 'onlineSavingsMsg'
Learn More

Di-Ammonium Hydrogen Citrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10418400 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10418400 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10418400 Supplier Fisher Chemical Supplier No. A/5200/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 3012-65-5
pH 4.3
Physical Form Solid
Quantity 500 g
Molecular Formula C6H14N2O7
MDL Number 13068
InChI Key YXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
IUPAC Name diammonium 3-carboxy-3-hydroxypentanedioate
Molecular Weight (g/mol) 226.19
Formula Weight 226.19g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid, diammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.