missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-iso-propyl-d14-amine HCl, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | N,N-Diisopropylamine-d14, Hydrochloride |
| CAS | 1219803-88-9 |
| Molecular Formula | C6 2H14 H N . Cl H |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C6H15N.ClH/c1-5(2)7-6(3)4;/h5-7 H,1-4H3;1 H/i1D3,2D3,3D3,4D3,5 D,6 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,3-heptadeuterio-N-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)propan-2-amine;hydrochloride |
| Molecular Weight (g/mol) | 151.74 |
| Formula Weight | 151.185 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction