Learn More
Di-n-decyl dimethylammonium bromide, 80% aq. gel
Product Code. 11319879
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. 11319879
Brand: Thermo Scientific Alfa Aesar B24832.36
Request Bulk or Custom Format
CAS: 2390-68-3 | C22H48BrN | 406.537 g/mol
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-n-decyl dimethylammonium bromide is an efficient phase-transfer catalyst useful in many organic reactions. It is useful in many organic transformations such as isomerization, esterification, and alkylation among others.
Solubility
Soluble in methanol.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2390-68-3
Molecular Formula C22H48BrN
Molecular Weight (g/mol) 406.537
MDL Number MFCD00012194
InChI Key UMGXUWVIJIQANV-UHFFFAOYSA-M
Synonym didecyldimethylammonium bromide, di-n-decyldimethylammonium bromide, n-decyl-n,n-dimethyldecan-1-aminium bromide, ddab, didecyl dimethyl ammonium bromide, unii-72l098xl5y, 1-decanaminium, n-decyl-n,n-dimethyl-, bromide, didecyldimethylamine, bromide, didecyl-dimethyl-ammonium bromide, didecyl dimethyl azanium bromide
PubChem CID 16957
IUPAC Name didecyl(dimethyl)azanium;bromide
SMILES CCCCCCCCCC[N+](C)(C)CCCCCCCCCC.[Br-]
Specifications
Linear Formula [CH3(CH2)9]2N(CH3)2Br
Refractive Index 1.457
Quantity 500 g
Solubility Information Soluble in methanol.
Formula Weight 406.54
Chemical Name or Material Di-n-decyl dimethylammonium bromide, 80% aq. gel
compliance-icons
Product Identifier
  • Di-n-decyldimethylammonium bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Didecyldimethylammonium bromide