missing translation for 'onlineSavingsMsg'
Learn More

Di-t-butyl N,N-Diisopropylphosphoramidite, TRC

Product Code. 30378827
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378827 Supplier Toronto Research Chemicals Supplier No. TRCD4294001G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Di-t-butyl N,N-Diisopropylphosphoramidite
CAS 137348-86-8
Molecular Formula C14 H32 N O2 P
Quantity 1 g
Synonym Phosphoramidous acid, N,N-bis(1-methylethyl)-, bis(1,1-dimethylethyl) ester, Phosphoramidous acid, bis(1-methylethyl)-, bis(1,1-dimethylethyl) ester (9CI), Di-tert-butyl N,N-diisopropylphosphoramidite, Di-tert-butyl diisopropylphosphoramidite, N,N-Diisopropyl-di-tert-butylphosphoramidite
InChI Formula InChI=1S/C14H32NO2P/c1-11(2)15(12(3)4)18(16-13(5,6)7)17-14(8,9)10/h11-12H,1-10H3
SMILES CC(C)N(C(C)C)P(OC(C)(C)C)OC(C)(C)C
IUPAC Name N-[bis[(2-methylpropan-2-yl)oxy]phosphanyl]-N-propan-2-ylpropan-2-amine
Molecular Weight (g/mol) 277.38
Formula Weight 277.2171
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.