Läs mer
Läs mer
MP Biomedicals Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) (MPQuanti™ only)
Handla allt MP Biomedicals ProdukterBeskrivning
- Diagnostic Kit for 25-Hydroxyvitamin D (25-OH-VD) (Serum/Plasma) is based on fluorescence immunoassay technology.
- The sample moves through the strip from sample pad to absorbent pad.
- Vitamin D in the sample will compete with the VD-BSA antigen coated on the membrane.
- The less Vitamin D in the sample, the more fluorescent microspheres conjugated with anti-VD antibodies can be captured by the VD-BSA antigen coated on the membrane (Test line).
- The concentration of Vitamin D in the sample is inversely related to the intensity of the fluorescent signal captured on the T line.
- According to the fluorescence intensity of the test and the standard curve, the concentration of Vitamin D in the sample is calculated by the MPQuanti™ Immuflorescence Analyzer to show Vitamin D concentration in the sample.
- CE marked
- For professional in-vitro diagnostic use
- Working only with MPQuanti™ Immunofluorescence Analyzer
- Linear working range: 5 -100 ng/mL, (R2) is 0.956
- Test results should be interpreted at 15 minutes
- Used as an aid to assessment of Vitamin D levels
Specifikationer
Specifikationer
| Detectable Analytes | 25-OH-Vitamin D |
| Detection Method | Immunofluorescence |
| Sample Type | Human serum, Human plasma |
| Type | POC, Quantitative, vitamin D, 25-OH-VD |
| Form | Ready-to-Use (RTU) |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?