Learn More
Diallyl phthalate, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -70°C |
| Density | 1.12 |
| Boiling Point | 160°C to 163°C (4 mmHg) |
| Flash Point | 166°C (330°F) |
| Odor | Pungent, Makes Eyes Water |
| Refractive Index | 1.519 |
| Quantity | 500 g |
| UN Number | UN3082 |
| Beilstein | 1880877 |
| Formula Weight | 246.27 |
| Show More |
Safety and Handling
- Diallyl phthalate
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P262-Do not get in eyes, on skin, or on clothing.
- P273-Avoid release to the environment.
- P311-Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contain: Diallyl phthalate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.