missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diallyl Phthalate, TRC
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-allyl ester |
| CAS | 131-17-9 |
| Molecular Formula | C14 H14 O4 |
| Quantity | 10 g |
| Synonym | 1,2-Benzenedicarboxylic acid, 1,2-di-2-propen-1-yl ester (ACI), 1,2-Benzenedicarboxylic acid, di-2-propenyl ester (9CI), Phthalic acid, diallyl ester (6CI, 8CI), Allyl phthalate, Daiso DAP monomer, DAP, DAP Monomer, Dap Tohto DT 170, DAP-M, Dapon R, Dappu, DCP-A, Diallyl phthalate, DT 170, KY 5200, Monoplex DAP, NSC 7667 |
| InChI Formula | InChI=1S/C14H14O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h3-8H,1-2,9-10H2 |
| SMILES | C=CCOC(=O)c1ccccc1C(=O)OCC=C |
| IUPAC Name | bis(prop-2-enyl) benzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 246.26 |
| Formula Weight | 246.0892 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?