Learn More
Checkers Diamondback™ Tunnel Only
Beskrivelse
- Checkers Diamondback 45 in. hose bridge system provides protection for valuable cable and hose lines while ensuring a safer method of passage for vehicles
- They replace wooden and metal fabricated ramps and can be used to protect lines up to 45 inches in diameter
- High density, all-weather polyurethane construction protects from damage by physical pressure and reduces cable and hose maintenance and replacement costs
- The lightweight, modular design of interlocking sections provides multi-length flexibility
- Checkers Hose Bridge Systems are intended for temporary installations only
Tekniske data
Tekniske data
| Dimensions (L x W x H) | 41 x 25 x 14 cm |
| Color | Yellow |
| Material | Steel |
| Type | Tunnel |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?