missing translation for 'onlineSavingsMsg'
Learn More

Diazenedicarboxamide, Dr. Ehrenstorfer

Product Code. 30324148
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324148 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324148 Supplier Dr. Ehrenstorfer Supplier No. DREC12209000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azodicarboxamide
CAS 123-77-3
Analyte or Component Names Azodicarboxamide
Shipping Condition Room Temperature
Molecular Formula C2 H4 N4 O2
Certifications/Compliance ISO/IEC 17025
Synonym Diazenedicarboxamide, 1,1'-Azobisformamide, 1,1'-Azobis[formamide], 1,1'-Azobiscarbamide, 1,1'-Azodiformamide, 1500TID, A 9660, AA 110S, ABFA, AC, AC (foaming agent), AC 1, AC 1000, AC 1000 (blowing agent), AC 135, AC 150P, AC 170, AC 1C, AC 1L, AC 2F-K3, AC 3, AC 3000, AC 3000H, AC 3C2, AC 3K2, AC 5000, AC 5000 (blowing agent), AC 902, AC-EFS, AC-K 3, AC-K-F 3, AC-LQ, AC-R, ADC, ADC 21, ADCA, ADK Stab OF 14, AK 2, AW 9, AZ 120, AZ 130, AZ 199, AZ 3050I, AZ 605, AZ Ultra 3050, AZ-H, AZ-HM, AZ-L 25, AZ-VI 25, AZ-VI 8, AZH 25, AZM 2S, AZO AZ 130, Azobis 50C, Azobis CA, Azobis CA 110B, Azobis CA 51C, Azobiscarbonamide, Azobiscarboxamide, Azobul, Azobul B, Azocel, Azodicarbamide, Azodicarbonamide, Azodicarboxylic acid diamide, Azodiformamide, Azoform A, Azoplastone, BX 81, BX 81 (blowing agent), CA 500, CS 4M, Cel-Span 693K, Cellborn DW 6, Cellcom A, Cellcom AC 7000DB, Cellmic 223, Cellmic 523, Cellmic C, Cellmic C 1, Cellmic C 121, Cellmic C 191, Cellmic C 2, Cellmic C 217, Cellmic C 22, Cellmic CA 500, Cellmic CAP, Cellmic CAP 500, Cellmic CE, Cellmic M 257, Cellmic MB 1031A, Cellmic MB 3013, Cellmic RUB, Celogen 125FF, Celogen 725B, Celogen 765A, Celogen AZ, Celogen AZ 120, Celogen AZ 130, Celogen AZ 150, Celogen AZ 199, Celogen AZ 3990, ChKhZ 21, ChKhZ 21r, D 1500, D 200, D 300LN, D 400, DP 18/47, DP 45/1, Daiblow AC, Daiblow AC 2040L, Emarcell BA, Evipor, Excellar S 10, FB 800, FBH 30, FE 788, Fascom AZ 4ED, Ferrocell AZC 13R, Ficel AC, Ficel EP-A, GMA 401, Genitron AC, Genitron AC 2, Genitron AC 3, Genitron AC 4, Genitron ACSP 4, Genitron EPC, HM 80A, Kempore, Kempore 125, Kempore 200, Kempore 60/40, Kempore R 125, Lagocell 20, Lucel ADA, Luvopor 9241, Luvopor ABF 70P-FF, MB 31, Microfine ADC 4075, NSC 41038, NSC 674447, Nortech MF 1062FA, Paramid K 1, Pinhole ACR 3, Pinhole AK 2, Poramid K 1, Porofor 505, Porofor ADC, Porofor ADC/E-C 2, Porofor ADC/L-C 2, Porofor ADC/M-C 1, Porofor ADC/S-C 2, Porofor ChKhZ 21, Porofor ChKhZ 21r, Porofor KA 9149, Porofor M-C 1, Rhenogran ADC/K 50, S 643, SO 20, SO-G 3, SO-G 3 (foamer), SOL, Supercell AZ 2D, Supercell K-D, Supercell L-D, Supercell V-D, Tracel DB 170, Tracel DB 201/50PE, Unicell D, Unicell D 1500, Unicell D 200, Unicell D 200A, Unicell D 400, Unicell DK 9, Unifoam 1100, Unifoam AZ, Unifoam AZ 40, Unifoam AZ 50, Unifoam AZ Ultra 3050I, Unifoam AZ Ultra 3220, Unifoam AZ Ultra 7043, Unifoam AZ-L, Unifoam AZ-L 25, Unifoam AZ-MFE 583, Unifoam AZ-T 8, Unifoam AZ-V 45, Unifoam AZ-VI 50, Unifoam AZH, Unifoam AZH 25, Unifoam AZH-M, Unifoam AZM, Unifoam AZS, Unifoam AZSO 20, Unifoam SOL, Unifoam Z, Uniform AZ Ultra 1035, Uquifoam L, VP 600, Vinfom AA 100, Vinfom AA 250H, Vinifor AC 3A, Vinifor AC-T, Vinyfor AC, Vinyfor AC 1, Vinyfor AC 1C, Vinyfor AC 1L, Vinyfor AC 2F, Vinyfor AC 3, Vinyfor AC 3C, Vinyfor AC 3C-K2, Vinyfor AC 3K2, Vinyfor AC 3K7, Vinyfor AC 3M, Vinyfor AC 3M-K2, Vinyfor AC 50S, Vinyfor AC-LQ, Vinyfor AC-R, Vinyfor AW 9, Vinyfor DW 6, Vinyfor FE 788, Vinyfor FZ 80, Vinyfor SE 30, Vinyfor ST 70, Vinyfor SW 7, Vinyfor SW 9, Vinylol AC, delta1,1'-Biurea
InChI Formula InChI=1S/C2H4N4O2/c3-1(7)5-6-2(4)8/h(H2,3,7)(H2,4,8)
Solution Type Neat
SMILES NC(=O)N=NC(=O)N
IUPAC Name (E)-carbamoyliminourea
Industry Type Environmental Testing
Molecular Weight (g/mol) 116.08
Quantity 100 mg
Formula Weight 116.0334 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.