missing translation for 'onlineSavingsMsg'
Learn More

Dibenzosuberenone, 97%

Product Code. 10675912 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675912 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10675912 Supplier Thermo Scientific Acros Supplier No. 112531000
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2222-33-5 | C15H10O | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2222-33-5
Melting Point 85.0°C to 91.0°C
Color White to Yellow or Beige
Loss on Drying 0.5% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C15H10O
MDL Number MFCD00003588
Quantity 100 g
Synonym dibenzosuberenone, 5-dibenzosuberenone, 5h-dibenzo a,d cyclohepten-5-one, 5h-dibenzo a,d 7 annulen-5-one, dibenzo a,d cyclohepten-5-one, 2,3:6,7-dibenzotropone, 2,3:6,7-dibenzosuberen-4-on-1, 5h-dibenzo a,d cycloheptenone
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,toluene,chloroform,and ethyl acetate,sparingly soluble in ethanol and methanol
InChI Key SNVTZAIYUGUKNI-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3C2=O
IUPAC Name dibenzo[1,2-a
Molecular Weight (g/mol) 206.24
PubChem CID 16679
Formula Weight 206.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Dibenzosuberenone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.