missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl N,N-Diisopropylphosphoramidite, TRC

Product Code. 30353398
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353398 1 g 1g
30365584 50 g 50g
30367891 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353398 Supplier Toronto Research Chemicals Supplier No. TRCD4175001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108549-23-1
Molecular Formula C20 H28 N O2 P
Molecular Weight (g/mol) 345.42
IUPAC Name N-bis(phenylmethoxy)phosphanyl-N-propan-2-ylpropan-2-amine
SMILES CC(C)N(C(C)C)P(OCc1ccccc1)OCc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H28NO2P/c1-17(2)21(18(3)4)24(22-15-19-11-7-5-8-12-19)23-16-20-13-9-6-10-14-20/h5-14,17-18H,15-16H2,1-4H3
Formula Weight 345.1858
Chemical Name or Material Dibenzyl N,N-Diisopropylphosphoramidite, Technical grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.