missing translation for 'onlineSavingsMsg'
Learn More

Dicarbonyl(2,4-pentanedionato)rhodium(I), 97%

Product Code. 11343698
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343698 1 g 1g
11480221 250 mg 250mg
11353698 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343698 Supplier Thermo Scientific Alfa Aesar Supplier No. 039295.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14874-82-9 | C7H7O4Rh | 258.03 g/mol

(Acetylacetonato)dicarbonylrhodium(I) is used in in situ formation of a floors-soluble hydroformylation catalyst of interest in molecular engineering. It is also used as a catalyst for various carbonylation reactions, silylcarbocyclizations, conjugate additions to eons, carbamoylstannation, and reduction of aromatic nitro compounds. It is also used in Hydroformylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14874-82-9
Molecular Formula C7H7O4Rh
Molecular Weight (g/mol) 258.03
MDL Number MFCD00009884
InChI Key BZCAWKOPWNIDOC-FGSKAQBVSA-M
Synonym (Acetylacetonato)dicarbonylrhodium(I), Acetylacetonatorhodium(I) dicarbonyl
IUPAC Name λ¹-rhodium(1+) bis(methanidylidyneoxidanium) (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].[C-]#[O+].[C-]#[O+].C\C([O-])=C\C(C)=O

Specifications

Melting Point 144°C to 147° (sublimation)
Color Green
Assay Percent Range 99%
Odor Odorless
Quantity 1 g
Solubility Information Soluble in acetone
Formula Weight 258.04
Physical Form Crystalline
Chemical Name or Material Dicarbonyl(2, 4-pentanedionato)rhodium(I)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.