missing translation for 'onlineSavingsMsg'
Learn More

Dichloralphenazone, TRC

Product Code. 30372352
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30372352 100 mg 100mg
30369883 25 mg -
30371863 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30372352 Supplier Toronto Research Chemicals Supplier No. TRCD431700100MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-30-8
Molecular Formula C11 H12 N2 O . 2 C2 H3 Cl3 O2
Molecular Weight (g/mol) 519.03
Synonym 3H-Pyrazol-3-one, 1,2-dihydro-1,5-dimethyl-2-phenyl-, compd. with 2,2,2-trichloro-1,1-ethanediol (1:2), Antipyrine, compd. with chloral hydrate (1:2) (6CI,8CI), 1,1-Ethanediol, 2,2,2-trichloro-, compd. with 1,2-dihydro-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one (2:1) (9CI), Chloral hydrate, compd. with antipyrine (2:1) (8CI), Bihypnal, Bonadorm, Dichloralantipyrin, Dichloralantipyrine, Dichloralphenazone, Dormuphar, Dormwell, Ibiodorm, Kloralfenazon, Sedor, Sominat, Welldorm
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one;2,2,2-trichloroethane-1,1-diol
SMILES CN1N(C(=O)C=C1C)c2ccccc2.OC(O)C(Cl)(Cl)Cl.OC(O)C(Cl)(Cl)Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12N2O.2C2H3Cl3O2/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10;2*3-2(4,5)1(6)7/h3-8H,1-2H3;2*1,6-7H
Formula Weight 515.9347
Chemical Name or Material Dichloralphenazone CIV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.