Learn More
Dichloro(p-cymene)ruthenium(II) Dimer, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 52462-29-0 |
| Molecular Formula | C20H28Cl4Ru2 |
| MDL Number | MFCD00064793 |
| Quantity | 25 g |
| SMILES | [Cl-][Ru+2]1([Cl-][Ru+2]23456(C7C2(=C3C4=C5(C6=7)C(C)C)C)([Cl-]1)[Cl-])12345C6C1(=C2C3=C4(C5=6)C(C)C)C |
| Molecular Weight (g/mol) | 612.39 |
| Percent Purity | 98% |
| Chemical Name or Material | Dichloro(p-cymene)ruthenium(II) Dimer |