missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethyl Methyl Ether, TRC

Product Code. 30377086
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377086 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377086 Supplier Toronto Research Chemicals Supplier No. TRCD4354405G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichloromethyl Methyl Ether
CAS 4885-02-3
Molecular Formula C2H4Cl2O
Quantity 5 g
Synonym 1,1-Dichlorodimethyl Ether, 1,1-Dichloromethyl Methyl Ether, Chloromyl, Dichloromethoxymethane, Dichloromethyl Methyl Ether, Methoxydichloromethane, Methyl alpha,alpha-dichloromethyl Ether, NSC 91480, alpha,alpha-Dichloromethyl Methyl Ether
InChI Formula InChI=1S/C2H4Cl2O/c1-5-2(3)4/h2H,1H3
SMILES COC(Cl)Cl
IUPAC Name dichloro(methoxy)methane
Molecular Weight (g/mol) 114.96
Formula Weight 113.9639
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.