missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethylphenylsilane, 98%

Product Code. 11304519
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304519 25 g 25g
11456896 100 g 100g
11466896 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304519 Supplier Thermo Scientific Alfa Aesar Supplier No. B23999.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 149-74-6 | C7H8Cl2Si | 191.13 g/mol

Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dichloromethylphenylsilane is used as a monomer (‘building block’) in the production of silicone polymers or silicone resins. Silicone polymers may be oils, greases and rubbers. It is also used as an intermediate (starting material) in the production of other organosilicon substances. It is a bifunctional reagent for the preparation of methylphenylsilyl derivatives of diols.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is sensitive to moisture. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-74-6
Molecular Formula C7H8Cl2Si
Molecular Weight (g/mol) 191.13
MDL Number MFCD00000490
InChI Key GNEPOXWQWFSSOU-UHFFFAOYSA-N
Synonym methylphenyldichlorosilane, dichloro methyl phenylsilane, phenylmethyldichlorosilane, silane, dichloromethylphenyl, dichlor-fenyl-methylsilane, dichloro methyl phenyl silane, dichloromethylphenyl silane, unii-9jr2a9v0ji, 9jr2a9v0ji, benzene, dichloromethylsilyl
PubChem CID 9006
IUPAC Name dichloro-methyl-phenylsilane
SMILES C[Si](Cl)(Cl)C1=CC=CC=C1

Specifications

Boiling Point 205°C to 206°C
Quantity 25 g
Linear Formula C6H5Si(CH3)Cl2
UN Number UN2437
Beilstein 970975
Flash Point 82°C (179°F)
Solubility Information Reacts with water.
Formula Weight 191.13
Percent Purity 98%
Odor Acrid
Refractive Index 1.519
Sensitivity Moisture sensitive
Density 1.173
Chemical Name or Material Dichloromethylphenylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.