missing translation for 'onlineSavingsMsg'
Learn More

Dichlorophene, TRC

Product Code. 30378428
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378428 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378428 Supplier Toronto Research Chemicals Supplier No. TRCD4359705G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dichlorophen
CAS 97-23-4
Molecular Formula C13 H10 Cl2 O2
Quantity 5 g
Synonym 2,2'-Methylenebis[4-chlorophenol], 2,2'-Dihydroxy-5,5'-dichlorodiphenylmethane, 4,4'-Dichloro-2,2'-methylenediphenol, 5,5'-Dichloro-2,2'-dihydroxydiphenylmethane, Anthiphen, Antifen, Bis(2-hydroxy-5-chlorophenyl)methane, Bis(5-chloro-2-hydroxyphenyl)methane, Bis(chlorohydroxyphenyl)methane, Cordocel, DDDM, DDM, Di(5-Chloro-2-hydroxyphenyl)methane, Dicestal, Dichlorofen, Dichlorophen, Dichlorophen B, Dichlorophene, Dichlorophene 10, Dichlorphen, Diclorofen, Didroxan, Didroxane, Difentan, Diphentane 70, Diphenthane 70, Embephen, Fungicide GM, Fungicide M, G 4, G 4 Pure, G 4 Technical, Gefir, Gingivit, Giv Gard G 4-40, Halenol, Hyosan, Korium, M 213, M 213 (biocide), NSC 38642, NSC 39467, Palacel, Panacide, Plath-Lyse, Prevental, Preventol, Preventol GD, Preventol GDC, Sandocide, Sindar G 4, Taeniatol, Teniathane, Teniatol, Teniotol, Vermithana, Xanbac
InChI Formula InChI=1S/C13H10Cl2O2/c14-10-1-3-12(16)8(6-10)5-9-7-11(15)2-4-13(9)17/h1-4,6-7,16-17H,5H2
SMILES Oc1ccc(Cl)cc1Cc2cc(Cl)ccc2O
IUPAC Name 4-chloro-2-[(5-chloro-2-hydroxyphenyl)methyl]phenol
Molecular Weight (g/mol) 269.12
Formula Weight 268.0058
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.