missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Carboxylic Acid (Diclofenac Metabolite), TRC

Product Code. 30179913
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179913 10 mg 10mg
30179771 25 mg 25mg
30179779 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179913 Supplier Toronto Research Chemicals Supplier No. TRCD43567010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13625-57-5
Molecular Formula C13 H9 Cl2 N O2
Molecular Weight (g/mol) 282.12
Synonym 2-(2,6-Dichlorophenylamino)benzoic Acid
IUPAC Name 2-(2,6-dichloroanilino)benzoic acid
SMILES OC(=O)c1ccccc1Nc2c(Cl)cccc2Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H9Cl2NO2/c14-9-5-3-6-10(15)12(9)16-11-7-2-1-4-8(11)13(17)18/h1-7,16H,(H,17,18)
Formula Weight 281.001
Chemical Name or Material 2-(2,6-Dichlorophenylamino)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.