missing translation for 'onlineSavingsMsg'
Learn More

Diclofenac Methyl Ester, TRC

Product Code. 30138269
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138269 250 mg 250mg
18031750 50 mg 50mg
18072840 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138269 Supplier Toronto Research Chemicals Supplier No. TRCD436500250MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15307-78-5
Molecular Formula C15 H13 Cl2 N O2
Molecular Weight (g/mol) 310.18
Synonym Acetic acid, [o-(2,6-dichloroanilino)phenyl]-, methyl ester (8CI), 2-[2-(2,6-Dichloroanilino)phenyl]acetate, Diclofenac methyl ester, Methyl 2-(2,6-dichloroanilino)phenylacetate, [o-(2,6-Dichloroanilino)phenyl]acetic acid methyl ester
IUPAC Name methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H13Cl2NO2/c1-20-14(19)9-10-5-2-3-8-13(10)18-15-11(16)6-4-7-12(15)17/h2-8,18H,9H2,1H3
Formula Weight 309.0323
Percent Purity >95%
Chemical Name or Material Diclofenac methyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.