missing translation for 'onlineSavingsMsg'
Learn More

Dicyclohexylmethane-4,4'-diisocyanate (Mixture of Isomers), >90%, TRC

Product Code. 30009795
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009795 50 g 50g
30010577 10 g 10g
18033250 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009795 Supplier Toronto Research Chemicals Supplier No. TRCD43928550G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5124-30-1
Molecular Formula C15 H22 N2 O2
Molecular Weight (g/mol) 262.35
Synonym 4,4'-Methylenebis(cyclohexyl isocyanate), 4,4'-Diisocyanatodicyclohexylmethane
IUPAC Name 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
SMILES O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C15H22N2O2/c18-10-16-14-5-1-12(2-6-14)9-13-3-7-15(8-4-13)17-11-19/h12-15H,1-9H2
Formula Weight 262.1681
Chemical Name or Material Dicyclohexylmethane-4,4'-diisocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.