missing translation for 'onlineSavingsMsg'
Learn More

Didecyl Phthalate (~90%), TRC

Product Code. 30381005
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381005 1 g
30370416 10 g
30369281 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381005 Supplier Toronto Research Chemicals Supplier No. TRCD4393951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-77-5
Molecular Formula C28 H46 O4
Molecular Weight (g/mol) 446.66
Synonym 1,2-Benzenedicarboxylic acid, 1,2-didecyl ester, 1,2-Benzenedicarboxylic acid, didecyl ester (9CI), Phthalic acid, didecyl ester (6CI,7CI,8CI), Bis(n-decyl)phthalate, DDP, Decyl phthalate, Di-n-decyl phthalate, Dicapryl phthalate, Didecyl phthalate, NSC 15319, Vinycizer 105, Phthalic acid didecyl ester
IUPAC Name didecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C28H46O4/c1-3-5-7-9-11-13-15-19-23-31-27(29)25-21-17-18-22-26(25)28(30)32-24-20-16-14-12-10-8-6-4-2/h17-18,21-22H,3-16,19-20,23-24H2,1-2H3
Formula Weight 446.3396
Chemical Name or Material Phthalic acid, bis-n-decyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.