missing translation for 'onlineSavingsMsg'
Learn More

Dideschloro Florfenicol, TRC

Product Code. 30240874
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240874 1 mg 1mg
30240724 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30240874 Supplier Toronto Research Chemicals Supplier No. TRCD4405001MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138872-76-1
Molecular Formula C12 H16 F N O4 S
Molecular Weight (g/mol) 289.32
IUPAC Name N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
SMILES CC(=O)N[C@H](CF)[C@H](O)c1ccc(cc1)S(=O)(=O)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H16FNO4S/c1-8(15)14-11(7-13)12(16)9-3-5-10(6-4-9)19(2,17)18/h3-6,11-12,16H,7H2,1-2H3,(H,14,15)/t11-,12-/m1/s1
Formula Weight 289.0784
Chemical Name or Material Dideschloro Florfenicol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.