missing translation for 'onlineSavingsMsg'
Learn More

Didesethyl Chloroquine, TRC

Product Code. 17924405
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17924405 10 mg 10mg
17904945 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17924405 Supplier Toronto Research Chemicals Supplier No. TRCD44096010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4298-14-0
Molecular Formula C14 H18 Cl N3
Molecular Weight (g/mol) 263.77
IUPAC Name 4-N-(7-chloroquinolin-4-yl)pentane-1,4-diamine
SMILES CC(CCCN)Nc1ccnc2cc(Cl)ccc12

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H18ClN3/c1-10(3-2-7-16)18-13-6-8-17-14-9-11(15)4-5-12(13)14/h4-6,8-10H,2-3,7,16H2,1H3,(H,17,18)
Formula Weight 263.1189
Percent Purity >95%
Chemical Name or Material Didesethylchloroquine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.