missing translation for 'onlineSavingsMsg'
Learn More

Didesmethyl Sibutramine Hydrochloride, TRC

Product Code. 30005764
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30005764 5 mg 5mg
30004020 10 mg 10mg
30003904 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005764 Supplier Toronto Research Chemicals Supplier No. TRCD4411705MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84484-78-6
Molecular Weight (g/mol) 288.26
SMILES Cl.CC(C)CC(N)C1(CCC1)c2ccc(Cl)cc2

Specifications

Quantity 5 mg
Molecular Formula C15H23Cl2N
Synonym (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-3-methylbutan-1-amine Hydrochloride, Sibutramine Hydrochloride Monohydrate Imp. G (Pharmeuropa) as Hydrochloride, N,N-Didesmethylsibutramine Hydrochloride, Didesmethylsibutramine Hydrochloride, Sibutramine Hydrochloride Monohydrate Impurity G, Sibutramine Impurity G
Percent Purity >95%
Chemical Name or Material Didesmethyl Sibutramine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.