missing translation for 'onlineSavingsMsg'
Learn More

Didesmethyl Sibutramine, TRC

Product Code. 30008848
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008848 50 mg 50mg
17953415 10 mg 10mg
17991195 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008848 Supplier Toronto Research Chemicals Supplier No. TRCD44116550MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84467-54-9
Molecular Formula C15 H22 Cl N
Molecular Weight (g/mol) 251.79
Synonym N,N-Didesmethylsibutramine, 1-(4-Chlorophenyl)-α-(2-methylpropyl)cyclobutanemethanamine, (±)-Didesmethylsibutramine, 1-[1-(4-Chlorophenyl)cyclobutyl]-3-methylbutan-1-amine, 1-[1-(4-Chlorophenyl)cyclobutyl]-3-methylbutylamine, N-Didemethylsibutramine, N-Didesmethylsibutramine
IUPAC Name 1-[1-(4-chlorophenyl)cyclobutyl]-3-methylbutan-1-amine
SMILES CC(C)CC(N)C1(CCC1)c2ccc(Cl)cc2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H22ClN/c1-11(2)10-14(17)15(8-3-9-15)12-4-6-13(16)7-5-12/h4-7,11,14H,3,8-10,17H2,1-2H3
Formula Weight 251.1441
Percent Purity >95%
Chemical Name or Material Didesmethyl Sibutramine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.