missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Bromomalonate, TRC

Product Code. 30374635
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374635 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374635 Supplier Toronto Research Chemicals Supplier No. TRCD44384025G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Bromomalonate
CAS 685-87-0
Molecular Formula C7H11BrO4
Quantity 25 g
Synonym Bromomalonic Acid Diethyl Ester, 2-Bromomalonic Acid Diethyl Ester, Diethyl 2-bromo-1,3-propanedioate, Diethyl 2-Bromomalonate, Diethyl 2-bromopropanedioate, Diethyl Bromomalonate, Diethyl α-Bromomalonate, Ethyl Bromomalonate, NSC 1985, α-Bromomalonic Ester
InChI Formula InChI=1S/C7H11BrO4/c1-3-11-6(9)5(8)7(10)12-4-2/h5H,3-4H2,1-2H3
SMILES CCOC(=O)C(Br)C(=O)OCC
Molecular Weight (g/mol) 239.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.