missing translation for 'onlineSavingsMsg'
Learn More

Diethyl difluoromalonate, 97%

Product Code. 11332877
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332877 1 g 1g
11342877 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332877 Supplier Thermo Scientific Alfa Aesar Supplier No. L16042.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 680-65-9 | C7H10F2O4 | 196.15 g/mol

Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl difluoromalonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680-65-9
Molecular Formula C7H10F2O4
Molecular Weight (g/mol) 196.15
MDL Number MFCD00221617
InChI Key WCRVWLHTROHXBB-UHFFFAOYSA-N
Synonym diethyl 2,2-difluoromalonate, diethyl difluoromalonate, 2,2-difluoro-malonic acid diethyl ester, 1,3-diethyl 2,2-difluoropropanedioate, difluoromalonic acid diethyl ester, diethyl2,2-difluoromalonate, propanedioic acid, 2,2-difluoro-, 1,3-diethyl ester, propanedioic acid, difluoro-, diethyl ester, diethyl 2,2-difluoromalonate #, diethyl2,2-difluoropropanedioatev
PubChem CID 260648
IUPAC Name diethyl 2,2-difluoropropanedioate
SMILES CCOC(=O)C(F)(F)C(=O)OCC

Specifications

Density 1.179
Boiling Point 94°C to 95°C (23 mmHg)
Refractive Index 1.38
Quantity 1 g
Beilstein 1783527
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 196.15
Percent Purity 97%
Chemical Name or Material Diethyl difluoromalonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.