missing translation for 'onlineSavingsMsg'
Learn More

Diethyl ether, Spectrophotometric Grade, 99+%, inhibitor free, Thermo Scientific™

Solvent For U.V. Spectrophotometric and General Use

Brand:  Thermo Scientific Alfa Aesar 040976.M1

Quantity 4 x 1L

 View more versions of this product

Product Code. 15476345

  • 264.00€ / 4 L
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

60-29-7
74.12
RTZKZFJDLAIYFH-UHFFFAOYSA-N
3283
ethoxyethane
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
CHEBI:35702
CCOCC

Specifications

60-29-7
0.713
-45°C (-49°F)
Sweet
>99%
586.3 hPa (440mm Hg) at 20°C
1155
Immiscible with water,miscible with alcohol.
CCOCC
74.12
CHEBI:35702
Spectrophotometric
Colorless liquid
4 x 1L
170°C (338°F)
1.3525
C4H10O
MFCD00011646
diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
RTZKZFJDLAIYFH-UHFFFAOYSA-N
ethoxyethane
3283
74.12
Diethyl ether
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.