missing translation for 'onlineSavingsMsg'
Learn More

Diethyl Ketomalonate, TRC

Product Code. 30381933
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30381933 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381933 Supplier Toronto Research Chemicals Supplier No. TRCD476633500MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl Ketomalonate
CAS 609-09-6
Molecular Formula C7H10O5
Quantity 500 mg
Synonym 2-Oxomalonic Acid Diethyl Ester, Carbonyl Diethyl Malonate, Diethyl 2-Oxomalonate, Diethyl 2-Oxopropane-1,3-dioate, Diethyl Mesoxalate, Diethyl Oxomalonate, Diethyl Oxopropanedioate, Ethoxycarbonyl Ethoxycarbonyl Ketone, Ethyl Ketomalonate, Ethyl Mesoxalate, Ethyl Oxomalonate, NSC 53478, Oxopropanedioic Acid Diethyl Ester
InChI Formula InChI=1S/C7H10O5/c1-3-11-6(9)5(8)7(10)12-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)C(=O)C(=O)OCC
IUPAC Name diethyl 2-oxopropanedioate
Molecular Weight (g/mol) 174.15
Formula Weight 174.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.