missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Diethyl Ether, TRC

Product Code. 30373620
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30373620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373620 Supplier Toronto Research Chemicals Supplier No. TRCD4443355G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylene Glycol-Diethyl Ether
CAS 112-36-7
Molecular Formula C8 H18 O3
Quantity 5 g
Synonym Ether, bis(2-ethoxyethyl) (7CI,8CI), 1,1'-Oxybis[2-ethoxyethane], 1-Ethoxy-2-(2-ethoxyethoxy)ethane, 2-(2-Ethoxyethoxy)-1-ethoxyethane, 2-Ethoxyethyl ether, 3,6,9-Trioxaundecane, B 0767, Bis(2-ethoxyethyl) ether, DEDG, Diethyl Carbitol, Diethylene glycol diethyl ether, Ethanol, 2,2'-oxybis-, diethyl ether, Ethyldiglyme, Hisolve EDE, Diethylene Glycol-Diethyl Ether
InChI Formula InChI=1S/C8H18O3/c1-3-9-5-7-11-8-6-10-4-2/h3-8H2,1-2H3
SMILES CCOCCOCCOCC
IUPAC Name 1-ethoxy-2-(2-ethoxyethoxy)ethane
Molecular Weight (g/mol) 162.227
Formula Weight 162.1256
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.