missing translation for 'onlineSavingsMsg'
Learn More

Diethylenetriamine-N,N,N',N',N'-Pentaacetic Acid, MP BIOMEDICALS

Product Code. 11408111 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408111 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408111 Supplier MP Biomedicals Supplier No. 0215147883
  • ONLINE EXCLUSIVE PRICE
    92.70€
    80.85€ / 250g
    Save 11.85€
    12% Off
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Application Notes:

  • Diethylenetriamine-N,N,N',N',N'-pentaacetic Acid is used as a chelating agent.
  • Diethylenetriaminepentaacetic acid may be used to synthesize chitosan (CH)-gadolinium (Gd) diethylenetriaminepentaacetic acid (DTPA) conjugates.

Usage Statement:

  • Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Key Applications:

  • Chelating agent

Specifications

Quantity 250 g
Molecular Weight (g/mol) 393.35 g/mol
Percent Purity 90%
Physical Form Powder or Waxy Solid
compliance-icons compliance-icons
Product Identifier
  • Diethylenetriamine-N,N,N',N",N"-pentaacetic Acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H361d-Suspected of damaging the unborn child.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Diethylenetriamine-N,N,N',N",N"-pentaacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.