Learn More
Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals
Product Code. 16657052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16657052 5 g 5g
16667052 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16657052 Supplier Thermo Scientific Acros Supplier No. 465920050
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-11-9
Molecular Formula C5H10ClNS
Molecular Weight (g/mol) 151.65
MDL Number MFCD00022106
InChI Key HUUSTUALCPTCGJ-UHFFFAOYSA-N
IUPAC Name (chloromethanethioyl)diethylamine
SMILES CCN(CC)C(Cl)=S
Specifications
Alpha Vector DIETHYLCARBAMOTHIOICCHLORIDE
Melting Point 45°C to 50°C
Color White to Yellow
Boiling Point 113°C
Flash Point 113°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 5 g
UN Number 3261
Sensitivity Moisture sensitive
Formula Weight 151.66
Physical Form Solid
Chemical Name or Material Diethylthiocarbamoyl chloride